Learn More
Zorifertinib, MedChemExpress
Product Code. 16477618
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
Product Code. 16477618
Brand: Medchem Express HY18750
MedChemExpress Zorifertinib (AZD3759) is a potent, orally active, central nervous system-penetrant, EGFR inhibitor. At Km ATP concentrations, the IC50s are 0.3, 0.2, and 0.2 nM for EGFRwt, EGFRL858R, and EGFRexon 19Del, respectively.
Zorifertinib (AZD3759) is a potent, orally active, central nervous system-penetrant, EGFR inhibitor. At Km ATP concentrations, the IC50s are 0.3, 0.2, and 0.2 nM for EGFRwt, EGFRL858R, and EGFRexon 19Del, respectively.
Specifications
Chemical Name or Material Zorifertinib
CAS 1626387-80-1
Color White
Purity Grade Notes Research
Molecular Formula C22H23ClFN5O3
Quantity 10 mg
Synonym AZD3759
Solubility Information DMSO : ≥ 50 mg/mL (108.72 mM)
SMILES ClC1=CC=CC(NC2=NC=NC3=CC(OC)=C(OC(N4[C@H](C)CN(C)CC4)=O)C=C23)=C1F
Molecular Weight (g/mol) 459.9
Formula Weight 459.9
Percent Purity 99.53%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease