missing translation for 'onlineSavingsMsg'
Learn More

ZNF92, Rabbit anti-Human, Polyclonal Antibody, Abnova™

Product Code. 16126839
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16126839 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16126839 Supplier Abnova Supplier No. H00168374D01P.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Sequence: MVDKTPVMCSHFAQDVWPEHSIKDSFQKVILRTYGKYGHENLQLRKDHKSVDACKVYKGGYNGLNQCLTTTDSKIFQCDKYVKVFHKFPNVNRNKIRHTGKKPFKCKNRGKSFCMLSQLTQHKKIHTREYSYKCEECGKAFNWSSTLTKHKIIHTGEKPYKCEECGKAFNRSSNLTKHKIIHTGEKPYKCEECGKAFNRSSTLTKHKRIHTEEKPYKCEECGKAFNQFSILNKHKRIHMEDKPYKCEECGKAFRVFSILKKHKIIHTGEKPYKCEECGKAFNQFSNLTKHKIIHTGEKPYKCDECGKAFNQSSTLTKHKRIHTGEKPYKCEECGKAFKQSSTLTEHKIIHTGEKPYKCEKCGKAFSWSSAFTKHKRNHMEDKPYKCEECGKAFSVFSTLTKHKIIHTREKPYKCEECGKAFNRSSIFTKHKIIHTEGKSYKCEKCGNAFNQSSNLTARKIIYTGEKPYKYEECDKAFNKFSTLITHQIIYTGEKPCKHECGRAFNKSSNYTKEKLQT

Specifications

Antigen ZNF92
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene zinc finger protein 92
Gene Accession No. BC036439.1
Gene Alias HPF12/HTF12/TF12
Gene Symbols ZNF92
Host Species Rabbit
Immunogen ZNF92 (AAH36439.1, 1 a.a. ∼ 517 a.a) full-length human protein.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 168374
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.