Learn More
ZNF597 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
483.00 EUR
Specifications
| Antigen | ZNF597 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
ZNF597 Polyclonal specifically detects ZNF597 in Human samples. It is validated for Western Blot.Specifications
| ZNF597 | |
| Polyclonal | |
| Rabbit | |
| NP_689670 | |
| 146434 | |
| Synthetic peptide directed towards the middle region of human ZNF597. Peptide sequence: GLAQHQKSHSAENTYESTNCDKHFNEKPNLALPEETFVSGPQYQHTKCMK | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| FLJ33071, zinc finger protein 597 | |
| ZNF597 | |
| IgG | |
| 48 kDa |