missing translation for 'onlineSavingsMsg'
Learn More

ZNF548 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

483.00 EUR

Specifications

Antigen ZNF548
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18289555
View Documents
Novus Biologicals
NBP1-80170
100 μL
483.00 EUR
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ZNF548 Polyclonal specifically detects ZNF548 in Human samples. It is validated for Western Blot.
Specifications

Specifications

ZNF548
Polyclonal
Rabbit
Human
FLJ32932, zinc finger protein 548
ZNF548
IgG
63 kDa
Western Blot
Unconjugated
RUO
NP_690873
147694
Synthetic peptide directed towards the N terminal of human ZNF548. Peptide sequence VHMAEEIFTCMEGWKDLPATSCLLQHQGPQSEWKPYRDTEDREAFQTGQN.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.