missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZNF367 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16571082
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16571082 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16571082 Supplier Invitrogen™ Supplier No. PA598259

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

ZNF367 Polyclonal Antibody for Western Blot, IHC (P), ELISA

Transcriptional activator. Isoform 1 may be involved in transcriptional activation of erythroid genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZNF367
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 3.81 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene ZNF367
Gene Accession No. Q0VDT2, Q7RTV3
Gene Alias 8030486J21Rik; AFF29; C2H2 zinc finger protein ZFF29; C2H2 zinc finger protein ZFF29a; CDC14 cell division cycle 14 homolog B; CDC14B; habp4; hyaluronan binding protein 4; hypothetical protein LOC550421; LOW QUALITY PROTEIN: zinc finger protein 367; ZFF29; ZFF29a; ZFF29b; Zfp367; zgc:112157; Zinc finger protein 367; Znf367
Gene Symbols Zfp367, ZNF367
Host Species Rabbit
Immunogen Recombinant Human Zinc finger protein 367 protein (83-151AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 195828, 238673
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.