missing translation for 'onlineSavingsMsg'
Learn More

ZM-447439, MedChemExpress

Product Code. 16436047
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16436047

Brand: Medchem Express HY10128

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ZM-447439 is an aurora kinase inhibitor with IC50s of 110 and 130 nM for aurora A and B, respectively.

ZM-447439 is an aurora kinase inhibitor with IC50s of 110 and 130 nM for aurora A and B, respectively.

Specifications

Chemical Name or Material ZM-447439
CAS 331771-20-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C29H31N5O4
Quantity 5 mg
Solubility Information DMSO : 25 mg/mL (48.68 mM; Need ultrasonic)
SMILES O=C(NC1=CC=C(NC2=C(C(C=C3OCCCN4CCOCC4)=NC=N2)C=C3OC)C=C1)C5=CC=CC=C5
Molecular Weight (g/mol) 513.59
Formula Weight 513.59
Percent Purity 98.16%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.