missing translation for 'onlineSavingsMsg'
Learn More

ZKSCAN1 Monoclonal Antibody (OTI3C4), TrueMAB™, OriGene

Product Code. p-200015945
Encompass_Preferred
Change view
Click to view available options
Quantity:
30 μL
100 μL
Unit Size:
100µL
30µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17514524 30 μL 30µL
17594514 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17514524 Supplier Origene Technologies Supplier No. TA810967S

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZKSCAN1 Monoclonal antibody specifically detects ZKSCAN1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen ZKSCAN1
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI3C4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ZKSCAN1
Gene Accession No. P17029
Gene Alias 5930429A01Rik; 9130423L19Rik; 9230118B16Rik; AI646829; AI788588; C87323; KOX18; LOC100720521; PHZ-37; zinc finger and SCAN domain containing 21; zinc finger and SCAN domain-containing protein 21; zinc finger protein 139; Zinc finger protein 36; zinc finger protein 36 (KOX 18); zinc finger protein KOX18; Zinc finger protein with KRAB and SCAN domains 1; zinc finger with KRAB and SCAN domains 1; Zkscan1; ZNF139; ZNF36; Zscan21; ZSCAN33
Host Species Mouse
Immunogen Full length human recombinant protein of human ZKSCAN1 produced in E.coli.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7586
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.