missing translation for 'onlineSavingsMsg'
Learn More

ZK824859 hydrochloride, MedChemExpress

Product Code. 16494808
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494808 10 mg 10mg
16477557 5 mg 5mg
16465606 100 mg 100mg
16421077 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16494808 Supplier Medchem Express Supplier No. HY114330A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress ZK824859 hydrochloride is an oral available and selective urokinase plasminogen activator (uPA) inhibitor with IC50s of 79 nM, 1580 nM and 1330 nM for human uPA, tPA, and plasmin, respectively.

ZK824859 hydrochloride is an oral available and selective urokinase plasminogen activator (uPA) inhibitor with IC50s of 79 nM, 1580 nM and 1330 nM for human uPA, tPA, and plasmin, respectively.

Specifications

Chemical Name or Material ZK824859 hydrochloride
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C23H23ClF2N2O4
Quantity 10 mg
Solubility Information DMSO : ≥ 125 mg/mL (268.88 mM)
SMILES CC1=CC(OC2=C(F)C=C(F)C(O[C@H](CC)C(O)=O)=N2)=CC(C3=CC=CC(CN)=C3)=C1.Cl
Molecular Weight (g/mol) 464.89
Formula Weight 464.89
Percent Purity 98.77%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.