Learn More
Zirconium ICP Standard, 10000 mg/L Zr in HCl 5%, HF 2%, Honeywell Fluka™
Shop All Honeywell Fluka ProductsDescription
Honeywell Fluka™ analytical standards offer a growing range of high quality inorganic and organic reference materials for the European market produced according to ISO 9001. The products are suitable for a wide range of industries and applications and come complete with a detailed Certificate of Analysis (CoA) in every delivery, and are eligible for free expert technical support ensuring you have the information you need to quickly start your analysis and pass an audit.
Specifications
Specifications
| Chemical Name or Material | Zirconium ICP Standard |
| pH | <2 |
| Quantity | 100 mL |
| Grade | Standards for ICP and ICP-MS |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.