Learn More
Ziprasidone, Lipomed
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
Description
Ziprasidone
Specifications
Specifications
| Chemical Name or Material | Ziprasidone n 1,2-Dimethoxyethane |
| CAS | 146939-27-7 |
| Analyte or Component Names | Ziprasidone n 1,2-Dimethoxyethane |
| Shipping Condition | Room Temperature |
| Molecular Formula | C21 H21 Cl N4 O S |
| Synonym | Ziprasidone |
| InChI Formula | InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H |
| SMILES | Clc1cc2NC(=O)Cc2cc1CCN3CCN(CC3)c4nsc5ccccc45 |
| IUPAC Name | 5-[2-[4-(1,2-benzothiazol-3-yl)piperazin-1-yl]ethyl]-6-chloro-1,3-dihydroindol-2-one |
| Industry Type | Pharmaceutical |
| Show More |