Learn More
Ziprasidone, Lipomed
Product Code. 30459011
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
Product Code. 30459011
Brand: Lipomed LPMZPR1234FB10
Lipomed delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose Lipomed for confidence, compliance, and analytical excellence.
Ziprasidone
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Ziprasidone n 1,2-Dimethoxyethane
CAS 146939-27-7
Analyte or Component Names Ziprasidone n 1,2-Dimethoxyethane
Shipping Condition Room Temperature
Molecular Formula C21 H21 Cl N4 O S
Synonym Ziprasidone
InChI Formula InChI=1 S/C16H21NO2.ClH/c1-12(2)17-10-14(18)11-19-16-9-5-7-13-6-3-4-8-15(13)16;/h3-9,12,14,17-18 H,10-11H2,1-2H3;1 H
SMILES Clc1cc2NC(=O)Cc2cc1CCN3CCN(CC3)c4nsc5ccccc45
IUPAC Name 5-[2-[4-(1,2-benzothiazol-3-yl)piperazin-1-yl]ethyl]-6-chloro-1,3-dihydroindol-2-one
Industry Type Pharmaceutical
Molecular Weight (g/mol) 412.94
Quantity 10 mg
Formula Weight 412.1125 g/mol