missing translation for 'onlineSavingsMsg'
Learn More

Zinc sulfate monohydrate, Zn 35.5%

Product Code. p-7049365
Click to view available options
Quantity:
1 kg
5 kg
Unit Size:
1kg
5kg
This item is not returnable. View return policy

Product Code. 11337757

Brand: Thermo Scientific Alfa Aesar 014160.A1

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7446-19-7 | H2O5SZn | 179.451 g/mol

Zinc sulfate monohydrate is used in calico-printing, preserving wood and skins, electrolytes for zinc plating, bleaching paper and clarifying glue. It is also used as a chemical reagent, coagulant in the preparation of rayon, mordant in dyeing and as a zinc source in animal feeds. In medicine, it is used as an astringent and emetic. Further, it is a precursor to the pigment lithopone. In addition to this, it is used to supply zinc in fertilizers, agricultural sprays, in electrolytes for zinc plating and as a mordant in dyeing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7446-19-7
Molecular Formula H2O5SZn
Molecular Weight (g/mol) 179.451
MDL Number MFCD00149893
InChI Key RNZCSKGULNFAMC-UHFFFAOYSA-L
Synonym zinc sulfate monohydrate, zinc sulfate hydrate, unii-ptx099xsf1, ptx099xsf1, sulfuric acid, zinc salt 1:1 , monohydrate, zinc sulphate monohydrate, gunningite, sulfato de zinc monohydratado, zinc sulfate tn, zinc sulfate,monohydrate
PubChem CID 62639
IUPAC Name zinc;sulfate;hydrate
SMILES O.[O-]S(=O)(=O)[O-].[Zn+2]

Specifications

Physical Form Coarse Powder
Quantity 1 kg
UN Number UN3077
Merck Index 14,10159
Solubility Information Soluble in water and alcohol.
Formula Weight 179.45 (161.43 Anhydrous)
Concentration or Composition (by Analyte or Components) Zn 35.5%
Chemical Name or Material Zinc sulfate monohydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.