missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZHX1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5113306
This item is not returnable.
View return policy
Description
ZHX1 Polyclonal Antibody for IHC (P), ELISA
The members of the zinc fingers and homeoboxes gene family are nuclear homodimeric transcriptional repressors that interact with the A subunit of nuclear factor-Y (NF-YA) and contain two C2H2-type zinc fingers and five homeobox DNA-binding domains. This gene encodes member 1 of this gene family. In addition to forming homodimers, this protein heterodimerizes with members 2 and 3 of the zinc fingers and homeoboxes family. Alternative splicing results in multiple transcript variants encoding the same protein.
Specifications
| ZHX1 | |
| Polyclonal | |
| Unconjugated | |
| ZHX1 | |
| Zhx1; zinc finger and homeodomain protein 1; zinc fingers and homeobox 1; zinc fingers and homeoboxes 1; zinc fingers and homeoboxes protein 1; zinc-fingers and homeoboxes 1 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 11244 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q9UKY1 | |
| ZHX1 | |
| Recombinant Human Zinc fingers and homeoboxes protein 1, isoform 2 protein (1-292AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction