Learn More
ZEOprep™ SH (Thiol) Functionalized
Product Code. 30048983 Shop All Zeochem Products
Change view
Click to view available options
:
1 kg
100 g
5 kg
500 g
Product Code.
30048983 100 g
30049030 1 kg
30049022 5 kg
30049005 500 g
4 options
Product Code. 30048983
Brand: ZEOprep 300177
ZEOprep™ is a packing material providing fast, effective, and reproducible separations. THIOL Irregular shaped silica gel is used in Gravity, Flash, and Medium Pressure Chromatography. The THIOL ligand makes an Ion Exchange material suitable for weak anion interactions.
  • SH (Thiol) functionalized for Ion Exchange separations
  • Idea for weak anion applications
  • Irregular Amorphous Silica Gel
  • High Purity
  • European Manufacture
  • Swiss accuracy and consistency
  • Excellent batch to batch reproducibility
  • ISO Certification
  • High Chemical and Mechanical Stability
  • Narrow Particles Size Distribution
  • 20 to 500 μm particles size range
  • Available in 60 A or 90 A pore size
  • High loadability
  • Selection of pack sizes available
  • Suitable for gravity, flash, prep and process scale
Specifications
Particle Size 40 to 63 μm
Pore Size 60 Å
Technique Chromatography
Quantity 100 g
Type Silica Gel
Material Silica Gel
Phase SH (Thiol) Functionalized
Particle Shape Irregular