Learn More
ZEObeads™
Click to view available options
Phase:
C18 Functionalized
C8 Functionalized
DIOL Functionalized
NH2 Functionalized
Quantity:
100 g
500 g
1 kg
5 kg
Unit Size:
100g
1kg
500g
5kg
Description
- Spheroidal Silica Gel
- High Purity
- Swiss accuracy and consistency
- Excellent batch to batch reproducibility
- ISO Certification
- Very wide portfolio
- Wide Selection of functionalization options
- High Chemical and mechanical stability
- 50 A pore size
- 20 to 30 μm particle size
- Suitable for medium to high pressure
- Ideal for flash applications
- High loadability
- Unsurpassed Service and Support
- Selection of pack sizes available
- Suitable for gravity, flash and process
Specifications
Specifications
| Particle Size | 20 to 30 μm |
| Pore Size | 50 Å |
| Technique | Chromatography |
| Quantity | 5 kg |
| Type | Silica Gel |
| Material | Silica Gel |
| Phase | DIOL Functionalized |
| Particle Shape | Spheroidal |