missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZDHHC5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA571409
This item is not returnable.
View return policy
Description
ZDHHC5 Polyclonal Antibody for Western Blot
Palmitoyl acyltransferase for the G-protein coupled receptor SSTR5. Also palmitoylates FLOT2. [UniProt].
Specifications
| ZDHHC5 | |
| Polyclonal | |
| Unconjugated | |
| Zdhhc5 | |
| 1110032A17Rik; AI451382; DHHC5; DHHC-5; Kiaa1748; membrane-associated DHHC5 zinc finger protein; palmitoyltransferase ZDHHC5; probable palmitoyltransferase ZDHHC5; ZDHHC5; Zinc finger DHHC domain-containing protein 5; zinc finger DHHC-type containing 5; zinc finger protein 375; zinc finger, DHHC domain containing 5; zinc finger, DHHC-type containing 5; Zisp; ZNF375 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 25921 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9C0B5 | |
| Zdhhc5 | |
| synthetic peptide directed towards the middle region of human ZDHHC5. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction