missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZDHHC5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA567578
This item is not returnable.
View return policy
Description
Predicted to react with Mouse and Rat samples.
Palmitoyl acyltransferase for the G-protein coupled receptor SSTR5. Also palmitoylates FLOT2. [UniProt].
Specifications
| ZDHHC5 | |
| Polyclonal | |
| Unconjugated | |
| Zdhhc5 | |
| 1110032A17Rik; AI451382; DHHC5; DHHC-5; Kiaa1748; membrane-associated DHHC5 zinc finger protein; palmitoyltransferase ZDHHC5; probable palmitoyltransferase ZDHHC5; ZDHHC5; Zinc finger DHHC domain-containing protein 5; zinc finger DHHC-type containing 5; zinc finger protein 375; zinc finger, DHHC domain containing 5; zinc finger, DHHC-type containing 5; Zisp; ZNF375 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 228136, 25921, 362156 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q2THW7, Q8VDZ4, Q9C0B5 | |
| Zdhhc5 | |
| Synthesized peptide derived from human ZDHHC5. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction