missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZCCHC3 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17781934
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17781934 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17781934 Supplier Invitrogen™ Supplier No. PA5120538

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: LO2, mouse liver. Immunogen sequence: SFRSAEKLAL FLRVYEEKRE QEDCWENFVV LGRSKSSLKT LFILFRNETV DVEDIVTWLK RHCDVLAVPV KVTDRFGIWT GEYKCEIELR QGEGGVRHLP GAFFLGAERG YSWYKGQPKT C.

Nucleic acid-binding protein involved in innate immune response to DNA and RNA viruses (PubMed:30193849, PubMed:30135424). Binds DNA and RNA in the cytoplasm and acts by promoting recognition of viral nucleic acids by virus sensors, such as DDX58/RIG-I, IFIH1/MDA5 and CGAS (PubMed:30193849, PubMed:30135424). Acts as a co-sensor for recognition of double-stranded DNA (dsDNA) by cGAS in the cytoplasm, thereby playing a role in innate immune response to cytosolic dsDNA and DNA virus (PubMed:30135424). Binds dsDNA and probably acts by promoting sensing of dsDNA by CGAS, leading to enhance CGAS oligomerization and activation (PubMed:30135424). Promotes sensing of viral RNA by RIG-I-like receptors proteins DDX58/RIG-I and IFIH1/MDA5 via two mechanisms: binds double-stranded RNA (dsRNA), enhancing the binding of DDX58/RIG-I and IFIH1/MDA5 to dsRNA and promotes 'Lys-63'-linked ubiquitination and subsequent activation of DDX58/RIG-I and IFIH1/MDA5 (PubMed:30193849). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZCCHC3
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.35 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ZCCHC3
Gene Accession No. Q9NUD5
Gene Alias C20orf99; ZCCHC3; Zinc finger CCHC domain-containing protein 3; zinc finger CCHC-type containing 3; zinc finger, CCHC domain containing 3
Gene Symbols ZCCHC3
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 215-335 of human ZCCHC3 (NP_149080.2).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 67917, 85364
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.