Learn More
ZC3H18 Rabbit anti-Human, Polyclonal, Proteintech
Product Code. 16814365 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
20 μL
150 μL
Unit Size:
150µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16814365 150 μL 150µL
16824365 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 16814365 Supplier Proteintech Supplier No. 258071AP150UL
Rabbit Polyclonal Antibody
ZC3H18 contains 1 C3H1-type zinc finger.
Specifications
Antigen ZC3H18
Applications Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene ZC3H18
Gene Accession No. Q86VM9
Gene Alias NHN1, Nuclear protein NHN1, ZC3H18
Gene Symbols ZC3H18
Host Species Rabbit
Immunogen ZC3H18 Fusion Protein Ag20481
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 124245
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG