missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZBTB1 Antibody, DyLight 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76970G
This item is not returnable.
View return policy
Description
ZBTB1 Polyclonal antibody specifically detects ZBTB1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| ZBTB1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| 22890 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 488 | |
| zinc finger and BTB domain containing 1 | |
| Antibody was raised against a 14 amino acid synthetic peptide near the center of human ZBTB1. The immunogen is located within amino acids 260 - 310 of ZBTB1. Amino Acid Squence: SAQTDKYRGDTSQA | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction