Learn More
Zardaverine, MedChemExpress
Product Code. 16409147
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
Product Code. 16409147
Brand: Medchem Express HY15485
MedChemExpress Zardaverine is a newly developed dual-selective PDE3/4 inhibitor with IC50 values of 0.5 uM and 0.8 uM respectively.
Zardaverine is a newly developed dual-selective PDE3/4 inhibitor with IC50 values of 0.5 uM and 0.8 uM respectively.
Specifications
Chemical Name or Material Zardaverine
CAS 101975-10-4
Color White
Purity Grade Notes Research
Molecular Formula C12H10F2N2O3
Quantity 5 mg
Solubility Information DMSO : 25 mg/mL (93.21 mM; Need ultrasonic)
SMILES O=C1C=CC(C2=CC=C(OC(F)F)C(OC)=C2)=NN1
Molecular Weight (g/mol) 268.22
Formula Weight 268.22
Percent Purity 98.16%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation