missing translation for 'onlineSavingsMsg'
Learn More

Zamicastat, MedChemExpress

Product Code. 16444068
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16444068 10 mg 10mg
16466767 5 mg 5mg
16435046 100 mg 100mg
16400427 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16444068 Supplier Medchem Express Supplier No. HY106004

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Zamicastat (BIA 5-1058) is a dopamine β-hydroxylase (DBH) inhibitor and can cross the blood-brain barrier (BBB) to cause central as well as peripheral effects. Zamicastat is also a concentration-dependent dual P-gp and BCRP inhibitor with IC50 values of 73.8 μM and 17.0 μM, respectively. Zamicastat reduces high blood pressure.

Zamicastat (BIA 5-1058) is a dopamine β-hydroxylase (DBH) inhibitor and can cross the blood-brain barrier (BBB) to cause central as well as peripheral effects. Zamicastat is also a concentration-dependent dual P-gp and BCRP inhibitor with IC50 values of 73.8 μM and 17.0 μM, respectively. Zamicastat reduces high blood pressure.

Specifications

Chemical Name or Material Zamicastat
CAS 1080028-80-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H21F2N3OS
Quantity 10 mg
Synonym BIA 5-1058
Solubility Information DMSO : 150 mg/mL (373.63 mM; Need ultrasonic)
SMILES S=C1N([C@@H]2CC3=CC(F)=CC(F)=C3OC2)C(CCNCC4=CC=CC=C4)=CN1
Molecular Weight (g/mol) 401.47
Formula Weight 401.47
Percent Purity 95.36%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.