missing translation for 'onlineSavingsMsg'
Learn More

(Z)-Thiothixene, MedChemExpress

Product Code. 16412986
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 16412986

Brand: Medchem Express HY108324

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (Z)-Thiothixene is an antagonist of serotonergic receptor extracted from patent US 20150141345 A1.

(Z)-Thiothixene is an antagonist of serotonergic receptor extracted from patent US 20150141345 A1.

Specifications

Chemical Name or Material (Z)-Thiothixene
CAS 3313-26-6
Color White
Purity Grade Notes Research
Molecular Formula C23H29N3O2S2
Quantity 25 mg
Solubility Information DMSO : 10 mg/mL (22.54 mM; ultrasonic and warming and heat to 60°C)
SMILES O=S(C1=CC(/C2=C\CCN3CCN(C)CC3)=C(SC4=C2C=CC=C4)C=C1)(N(C)C)=O
Molecular Weight (g/mol) 443.63
Formula Weight 443.63
Percent Purity 99.09%
Grade Research
Physical Form Powder
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.