Learn More
Z-Ile-Leu-aldehyde, MedChemExpress
Product Code. 16467907
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. 16467907
Brand: Medchem Express HY12465
MedChemExpress Z-Ile-Leu-aldehyde (Z-IL-CHO) is a potent and competitive peptide aldehyde inhibitor of γ-secretase and notch.
Z-Ile-Leu-aldehyde (Z-IL-CHO) is a potent and competitive peptide aldehyde inhibitor of γ-secretase and notch.
Specifications
Chemical Name or Material Z-Ile-Leu-aldehyde
Physical Form Solid
CAS 161710-10-7
Color White
Quantity 5 mg
Purity Grade Notes Research
Molecular Formula C20H30N2O4
For Use With (Application) Cancer-programmed cell death
Synonym Z-IL-CHO GSI-XII γ-Secretase inhibitor XII
Solubility Information DMSO : ≥ 41 mg/mL (113.12 mM)
SMILES O=C(OCC1=CC=CC=C1)N[C@H](C(N[C@H](C=O)CC(C)C)=O)[C@@H](C)CC
Molecular Weight (g/mol) 362.46
Formula Weight 362.46
Percent Purity 98.0%
Grade Research