missing translation for 'onlineSavingsMsg'
Learn More

(Z)-9-Propenyladenine, MedChemExpress

Product Code. 16464226
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
1mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16464226 100 mg 100mg
16482768 10 mg 10mg
16480499 1 mg 1mg
16475507 5 mg 5mg
16459466 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16464226 Supplier Medchem Express Supplier No. HY100079A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress (Z)-9-Propenyladenine is a mutagenic impurity in tenofovir disoproxil fumarate. Tenofovir is an antiretroviral drug known as nucleotide analogue reverse transcriptase (NtART) inhibitor, which blocks reverse transcriptase, a crucial virus enzyme in HIV-1 and HBV.

(Z)-9-Propenyladenine is a mutagenic impurity in tenofovir disoproxil fumarate. Tenofovir is an antiretroviral drug known as nucleotide analogue reverse transcriptase (NtART) inhibitor, which blocks reverse transcriptase, a crucial virus enzyme in HIV-1 and HBV.

Specifications

Chemical Name or Material (Z)-9-Propenyladenine
CAS 1464851-21-5
Color White
Purity Grade Notes Research
Molecular Formula C8H9N5
Quantity 100 mg
Synonym (Z)-Mutagenic Impurity of Tenofovir Disoproxil
Solubility Information DMSO : ≥ 100 mg/mL (570.81 mM)
SMILES NC1=C2N=CN(/C=C\C)C2=NC=N1
Molecular Weight (g/mol) 175.19
Formula Weight 175.19
Percent Purity 98.02%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-anti-virus
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.