missing translation for 'onlineSavingsMsg'
Learn More

Melford YPD, 1G Capsules

Product Code. 30018952
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
This item is not returnable. View return policy

Product Code. 30018952

Brand: Melford Y200851000.0

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mix 65 capsules in one liter distilled water until evenly dispersed.

Mix 65 capsules in one liter distilled water until evenly dispersed. Heat with repeated stirring and boil for one minute to dissolve completely. Distribute and autoclave at 121°C for 15 minutes.

Ingredients:

Peptone: 20 g/l

Dextrose: 20 g/l

Yeast Extract: 10 g/l

Agar: 15 g/l

Specifications

Quantity 1 kg
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.