missing translation for 'onlineSavingsMsg'
Learn More

YK-4-279, MedChemExpress

Product Code. p-200098489
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16492217 50 mg 50mg
16476476 100 mg 100mg
16406088 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16492217 Supplier Medchem Express Supplier No. HY14507

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress YK 4-279 is an inhibitor of RNA Helicase A (RHA) binding to the oncogenic transciption factor EWS-FLI1.

YK 4-279 is an inhibitor of RNA Helicase A (RHA) binding to the oncogenic transciption factor EWS-FLI1.

Specifications

Chemical Name or Material YK-4-279
CAS 1037184-44-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C17H13Cl2NO4
Quantity 50 mg
Solubility Information DMSO : 25 mg/mL (68.27 mM; Need ultrasonic)
SMILES ClC1=C2C(C(O)(CC(C3=CC=C(OC)C=C3)=O)C(N2)=O)=C(Cl)C=C1
Molecular Weight (g/mol) 366.2
Formula Weight 366.2
Percent Purity 99.61%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.