missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YBX1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20339-1-AP-20UL
This item is not returnable.
View return policy
Description
YBX1 binds to splice sites in pre-mRNA and regulates splice site selection. This protein binds and stabilizes cytoplasmic mRNA and contributes to the regulation of translation by modulating the interaction between the mRNA and eukaryotic initiation factors. It binds to promoters that contain a Y-box (5′-CTGATTGGCCAA-3′), such as HLA class II genes. It regulates the transcription of numerous genes and promotes separation of DNA strands that contain mismatches or are modified by cisplatin. It has endonucleolytic activity and can introduce nicks or breaks into double-stranded DNA (in vitro), and it may play a role in DNA repair.Specifications
| YBX1 | |
| Polyclonal | |
| Unconjugated | |
| YBX1 | |
| BP 8, CBF A, CSDA2, CSDB, DBPB, DNA binding protein B, EFI A, Enhancer factor I subunit A, MDR NF1, NSEP 1, NSEP1, Y box binding protein 1, Y box transcription factor, YB 1, YB1, YBX1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 22608, 4904 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunohistochemistry (Paraffin), Immunofluorescence, Immunoprecipitation, Western Blot | |
| 0.39 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P62960, P67809 | |
| YBX1 | |
| YBX1 Fusion Protein Ag14129 | |
| 20 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction