Learn More
YAP1 Mouse anti-Human, Unconjugated, Clone: 3A7A9, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66900-1-IG-20UL
 View more versions of this product
Product Code. 16809185
  • 145.00 EUR / 20µL
Description
The YAP1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human YAP1. This antibody recognizes human, mouse, rat antigen. The YAP1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
YAP1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunoprecipitation, Immunohistochemistry, Immunofluorescence
3A7A9
Unconjugated
20 μL
10413
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions