missing translation for 'onlineSavingsMsg'
Learn More
Learn More
YAP/TAZ inhibitor-1, MedChemExpress
Description
YAP/TAZ inhibitor-1 is a YAP/TAZ inhibitor extracted from patent WO2017058716A1, Compound 1, has an IC50 of <0.100 μμ in firefly luciferase assay.
Specifications
Specifications
| Chemical Name or Material | YAP/TAZ inhibitor-1 |
| CAS | 2093565-23-0 |
| Color | Light Yellow |
| Purity Grade Notes | Research |
| Molecular Formula | C33H39N3O5S2 |
| Quantity | 10 mg |
| Solubility Information | DMSO : 62.5 mg/mL (100.51 mM; Need ultrasonic) |
| SMILES | O=S(C1=CC2=C(C=C1)C3=C(C=C(S(=O)(C4CCC(C)(C)CC4)=O)C=C3)/C2=N\O)(N5CCN(C(C6=CC=CC=C6)C)CC5)=O |
| Molecular Weight (g/mol) | 621.81 |
| Formula Weight | 621.81 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?