missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Xylene, Mixed Isomers, Multisolvent™ Grade, Sharlab
Description
Multisolvent™ products are most frequently use for HPLC, but can also be used for numerous other applications, making it one of the most universal grades available. These versatile products have numerous advantages over application-specific solvents:
- Guarantee results: QC controls are performed by modern analytical techniques for all guaranteed applications
- Optimize purchasing and stocking: One product for most applications prevents the need to order, stock and track the supply of multiple grade
- Optimal packaging: Highly-resistant and durable amber glass bottles or stainless steel drums ensure the highest product safety and quality
- Lower costs by unifying: Multiple-use products at single-grade prices to avoid buying small quantities of specific products
- Preserve the environment: Returnable packaging (one-way glass bottles or SS returnable drums) help prevent waste
Specifications
Specifications
| Melting Point | -34°C |
| CAS | 1330-20-7 |
| Color | Colorless |
| Acidity | Max. 0.00025meq/g |
| Alkalinity | Max. 0.00025meq/g |
| Appearance | Clear |
| Density | 0.86g/cm3 |
| Quantity | 1 L |
| Flash Point | 25°C |
| Ignition Point | 465°C |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?