Learn More
Invitrogen™ XRCC1 Monoclonal Antibody (10E10)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA549160
Product Code. 17941481
  • 415.00 EUR / 100µg
Description
Adding 0.2 mL of distilled water will yield a concentration of 500 μg/mL. Positive Control - WB: human Hela whole cell, human 293T whole cell, human K562 whole cell, human Jurkat whole cell, human HEL whole cell, human A431 whole cell. ICC/IF: A431 cell. Flow: Hela cell. Store at -20°C for one year from date of receipt. After reconstitution, at 4°C for one month. It can also be aliquotted and stored frozen at -20°C for six months. Avoid repeated freeze-thaw cycles.
XRCC1 (X-ray cross complementing factor-1) is involved in DNA base excision repair. XRCC1 is a 70 kDa protein that has been shown to be physically associated with other DNA repair enzymes, including poly (ADP-ribose) polymerase (PARP), DNA Ligase III and DNA polymerase beta. XRCC1 contains a BRCT domain (for BRCA1 carboxyl terminus) which was originally found in the tumor suppressor protein BRCA1. Recently, the BRCT domain has been shown to mediate the protein-protein interaction of XRCC1 and DNA ligase III-alpha.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
XRCC1
Monoclonal
500 μg/mL
PBS with 4mg trehalose and no preservative
P18887
XRCC1
E. coli-derived human XRCC1 recombinant protein (Position: E538-A633).
100 μg
Primary
Human
Antibody
IgG2b
Flow Cytometry, Western Blot, Immunocytochemistry
10E10
Unconjugated
XRCC1
DNA repair protein XRCC1; RCC; X-ray repair complementing defective repair in Chinese hamster cells 1; X-ray repair cross complementing 1; x-ray repair cross-complementing group 1 protein; X-ray repair cross-complementing protein 1; XRCC1; Xrcc-1
Mouse
Antigen affinity chromatography
RUO
7515
-20°C
Lyophilized
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions