missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XMD-17-51 Trifluoroacetate, MedChemExpress
Description
XMD-17-51 Trifluoroacetate is a pyrimido-diazepinone compound that is able to modulate protein kinases.
Specifications
Specifications
| Chemical Name or Material | XMD-17-51 Trifluoroacetate |
| Color | Off-White |
| Purity Grade Notes | Research |
| Molecular Formula | C23H25F3N8O3 |
| Quantity | 5 mg |
| Solubility Information | DMSO : 100 mg/mL (192.87 mM; Need ultrasonic) |
| SMILES | O=C1N(C)C2=CN=C(NC3=CN(C4CCNCC4)N=C3)N=C2N(C)C5=CC=CC=C15.FC(C(O)=O)(F)F |
| Molecular Weight (g/mol) | 518.49 |
| Formula Weight | 518.49 |
| Percent Purity | 99.34% |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?