missing translation for 'onlineSavingsMsg'
Learn More

Scharlab XLD Agar

Product Code. 11476257
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11476257 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11476257 Supplier Scharlab Supplier No. 01211500

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Scharlab XLD Agar is used for the isolation of enteropathogenic species, especially Salmonella and Shigella.

  • AOAC/BAM/COMPF/EP/HP/ISO/USP
  • Solid culture medium for the isolation of enteropathogenic species, especially Salmonella and Shigella
  • According to Phamacopoeia Harmonised method and ISO 6340 standards.

Specifications

Certifications/Compliance ISO 6340
Color Red
Form Solid
Media Type XLD Agar
Product Type Differential Medium
Target Organism Class Gram-negative Microorganisms
Formulation Xylose 3.50 g/L, L-Lysine 5.00 g/L, Lactose 7.50 g/L, Sucrose 7.50 g/L, Sodium Chloride 5.00 g/L, Yeast Extract 3.00 g/L, Phenol Red 0.08 g/L, Sodium Deoxycholate 2.50 g/L, Sodium Thiosulfate 6.80 g/L, Ammonium Ferric Citrate 0.80 g/L, Agar 15.00 g/L
Incubation Conditions 30°C to 35°C
Incubation Time 18 to 48 hr.
pH 7.4 ±0.2
Quantity 500 g
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.