Learn More
Xanthine monosodium salt monohydrate, ≥96% (dry basis)
Product Code. 15875198 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
5 g
10 g
100 g
Packaging:
Plastic bottle
Unit Size:
100g
10g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15875198 5 g Plastic bottle 5g
15885198 10 g Plastic bottle 10g
15895198 100 g Plastic bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15875198 Supplier Thermo Scientific Supplier No. J23465.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
CAS: 1196-43-6 | C5H3N4NaO2 | 174.095 g/mol
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 1196-43-6
Molecular Formula C5H3N4NaO2
Molecular Weight (g/mol) 174.095
MDL Number MFCD00069669
InChI Key HKDXLLDCXWUUKY-UHFFFAOYSA-M
Synonym xanthine sodium salt, xanthine, sodium salt, xanthate de sodium french, 1 h-purine-2,6-dione, 3,7-dihydro-, sodium salt, 3,7-dihydro-1 h-purine-2,6-dione sodium salt, 3,7-dihydro-1h-purine-2,6-dione, sodium salt, xanthate de sodium, 1h-purine-2,6-dione, 3,9-dihydro-, sodium salt 1:?, potassium 2-oxo-3,7-dihydropurin-6-olate, sodium 2-oxo-3,7-dihydropurin-6-olate
PubChem CID 23668765
IUPAC Name sodium;2-oxo-3,7-dihydropurin-6-olate
SMILES C1=NC2=C(N1)C(=NC(=O)N2)[O-].[Na+]
Specifications
Physical Form Crystalline Powder
Quantity 5 g
Formula Weight 192.11
Percent Purity ≥96%
Packaging Plastic bottle
Assay Dry basis
Chemical Name or Material Xanthine Monosodium Salt Monohydrate