missing translation for 'onlineSavingsMsg'
Learn More
Learn More
XAB1 Antibody (GPN1/2350), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-75783AF750
This item is not returnable.
View return policy
Description
XAB1 Monoclonal specifically detects XAB1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| XAB1 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| GPN1 | |
| Recombinant full-length human XAB1 protein (Uniprot: Q9HN4) | |
| 0.1 mL | |
| Signal Transduction | |
| GPN1 is involved in protein synthesis events. It is expressed ubiquitously with highest expression in testis. It binds to the RNA polymerase II- (Poll II) associated proteins RPAP1-3 and to XPA (a protein involved in DNA repair mechanisms), thereby forming an interface with Poll II. Via this interaction, GPN1 is thought to mediate the involvement of Pol II in both protein complex formation and protein chaperone/ scaffolding activities. In addition, GPN1 interacts with components of the integrator and molecular chaperone complexes, further implicating GPN1 in protein assembly. GPN1 contains a cluster of acidic amino acids in its C-terminal region and a series of sequences similar to those found in GTP-binding proteins in its N-terminal region, suggesting that GPN1 has possible GTPase activity. | |
| Store at 4°C in the dark. |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| GPN1/2350 | |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| ATP(GTP)-binding protein, ATPBD1A, GPN-loop GTPase 1, GTPase, MBD2-interacting protein, MBDin, MBDINXPA binding protein 1, XAB1FLJ51176, XPA-binding protein 1 | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction