missing translation for 'onlineSavingsMsg'
Learn More

Tork Constant Air Freshener Refill A3
GREENER_CHOICE

Product Code. 30243724 Shop All Tork Products
Click to view available options
Scent:
Breeze
Floral
Lemongrass
Variety Pack
Unit Size:
Pack of 6
This item is not returnable. View return policy

Product Code. 30243724

Brand: Tork 257012

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Tork Constant Air Freshener help to keep washrooms constantly fresh with odor neutralizer.

  • For a fresh washroom at all times, this scent contains an odour neutraliser to eliminate smells, not just mask them.
  • This propellant-free system uses only pure perfume oil, isn't flammable, and is not pressurised.
  • Use with Tork Constant Air Freshener Dispensers for long-lasting, constant scent dispersal.

Refill packs available:

  • Blossom scent with delicate floral notes for an inviting feel.
  • Breeze scent with ylang-ylang, floral and woody notes for a fresh feel.
  • Odour neutraliser scent with a very subtle, unobtrusive lemongrass smell.
  • Variety pack includes 3 each of Blossom and Breeze.
TRUSTED_SUSTAINABILITY

Specifications

Product Type Air Freshener Refill
Alternate Part Number 7322541889351
Color-coded No
Form Spray
Scent Lemongrass
Quantity 6/Pk.
Volume (Metric) 32 mL
compliance-icons
Signal Word
  • Warning
Hazard Category
  • Skin sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.