Learn More
Value Plastics Female Luer, Panel Mount 1/4-28 UNF to 200 Series Barb
Product Code. 30054071 Shop All Cole Parmer Products
Change view
Click to view available options
Diameter (English) Inner:
1/16 in.
1/8 in.
3/32 in.
Material:
Nylon
Polycarbonate
Polypropylene
Unit Size:
Pack of 25
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (English) Inner Material unitSize
30054071 1/16 in. Polypropylene Pack of 25
30053104 1/8 in. Nylon Pack of 25
30053510 1/16 in. Polycarbonate Pack of 25
30053397 3/32 in. Polypropylene Pack of 25
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 30054071 Supplier Value Plastics Supplier No. 5012065
Only null left
Add to basket
Add to basket
Luer tapers that are constructed to meet ISO 594-1 and 594-2 standards
Rotating locks minimize twisting and kinking of tubing during positioning. Tapered cone seals are reliable and uncomplicated. Facilitate single-use configurations that minimize downtime
Specifications
Certifications/Compliance ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP
Diameter (English) Inner 1/16 in.
Material Polypropylene
Packaging Quantity 25/Pk.
Inlet Female Luer with Panel Mount, Hose Barb
Always test the application to verify the performance of the interconnection.