Description
Rotating locks minimize twisting and kinking of tubing during positioning. Tapered cone seals are reliable and uncomplicated. Facilitate single-use configurations that minimize downtime
Specifications
Specifications
| Certifications/Compliance | ADCF;FDA;Non-DEHP;Non-Phthalate;RoHS;USP |
| Diameter (English) Inner | 1/16 in. |
| Material | Polypropylene |
| Packaging Quantity | 25/Pk. |
| Inlet | Female Luer with Panel Mount, Hose Barb |
Always test the application to verify the performance of the interconnection.