Learn More
Fisherbrand X2 470mm Side Arm Shaft, for Wrist Motion Shaker
Brand:  Fisherbrand 88861194
X2 470mm Side Arm Shaft, for Wrist Motion Shaker
Product Code. 15554130
  • ONLINE EXCLUSIVE PRICE
    47.55 EUR
    45.20 EUR / Pack of 2
    Save 2.35 EUR
    4% Off
Only null left
Estimated Shipment: 24-04-2026