missing translation for 'onlineSavingsMsg'
Learn More

IKA X10 Disposable grinding chamber sterile, 40 ml

Brand:  IKA 0020001984

179.06 EUR valid until 2026-05-31
BEST PRICE on Promo! Use promo code "27998" to get your promotional price.


X10 Disposable grinding chamber sterile, 40 ml

Product Code. 15369768

  • 192.00 EUR / Pack of 10
Save Up to 70% on Equipment and Instruments
Use promo code 27998
View all promotions
Only null left
Estimated Shipment: 28-04-2026
Add to basket
This item is not returnable. View return policy

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.