Learn More
ABEBA™ X-Light White Occupational Shoes O2 ESD
Click to view available options
Size:
35
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Description
- Lightweight, comfortable occupational shoes
- Comply to EN ISO 20347: 2012, O2 SRC
- ESD dissipative according EN 61340 class climatic 1
- Slip resistant Mono PU outsole SRC
- White smooth leather
- Silver ions lining
- Replaceable active comfort insole
Specifications
Specifications
| Color | White |
| Material | Leather |
| Size | 41 |
| Certifications/Compliance (Europe) | Comply to EN 20347: 2012 O2 SRC, ESD dissipative according EN 61340 |
| Gender | Unisex |
| Ankle Style | Low-cut |
| Waterproof | Water Resistant |
| Product Type | O2 SRC ESD |
Product Content Correction
Product Title
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto