missing translation for 'onlineSavingsMsg'
Learn More

X-376, MedChemExpress

Product Code. 16497088
Encompass_Preferred
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
2mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16497088

Brand: Medchem Express HY16590

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress X-376 is a potent and highly specific ALK tyrosine kinase inhibitor (TKI) (IC50=0.61 nM). X-376 is a less potent inhibitor of MET (IC50=0.69 nM). X-376 displays potent anti-tumor activity.

X-376 is a potent and highly specific ALK tyrosine kinase inhibitor (TKI) (IC50=0.61 nM). X-376 is a less potent inhibitor of MET (IC50=0.69 nM). X-376 displays potent anti-tumor activity.

Specifications

Chemical Name or Material X-376
CAS 1365267-27-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C25H25Cl2FN6O3
Quantity 10 mg
Solubility Information DMSO : 100 mg/mL (182.68 mM; Need ultrasonic)
SMILES O=C(C1=NN=C(N)C(O[C@@H](C2=C(Cl)C=CC(F)=C2Cl)C)=C1)NC3=CC=C(C(N4CCN(C)CC4)=O)C=C3
Molecular Weight (g/mol) 547.41
Formula Weight 547.41
Percent Purity 98.37%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.