missing translation for 'onlineSavingsMsg'
Learn More

WZB117, MedChemExpress

Product Code. p-200100472
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16483707

Brand: Medchem Express HY19331

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress WZB117 is a glucose transporter 1 (Glut1) inhibitor, which downregulates glycolysis, induces cell-cycle arrest, and inhibits cancer cell growth in vitro and in vivo.

WZB117 is a glucose transporter 1 (Glut1) inhibitor, which downregulates glycolysis, induces cell-cycle arrest, and inhibits cancer cell growth in vitro and in vivo.

Specifications

Chemical Name or Material WZB117
CAS 1223397-11-2
Color Off-White
Purity Grade Notes Research
Molecular Formula C20H13FO6
Quantity 50 mg
Solubility Information DMSO : ≥ 150 mg/mL (407.27 mM)
SMILES OC1=CC=CC(C(OC2=CC=CC(F)=C2OC(C3=CC(O)=CC=C3)=O)=O)=C1
Molecular Weight (g/mol) 368.31
Formula Weight 368.31
Percent Purity 99.24%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.