missing translation for 'onlineSavingsMsg'
Learn More

WYE-687, MedChemExpress

Product Code. 16482517
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16482517 50 mg 50mg
16436706 100 mg 100mg
16416418 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16482517 Supplier Medchem Express Supplier No. HY15271

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress WYE-687 is an ATP-competitive mTOR inhibitor with an IC50 of 7 nM. WYE-687 concurrently inhibits activation of mTORC1 and mTORC2. WYE-687 also inhibits PI3Kα and PI3Kγ with IC50s of 81 nM and 3.11 μM, respectively.

WYE-687 is an ATP-competitive mTOR inhibitor with an IC50 of 7 nM. WYE-687 concurrently inhibits activation of mTORC1 and mTORC2. WYE-687 also inhibits PI3Kα and PI3Kγ with IC50s of 81 nM and 3.11 μM, respectively.

Specifications

Chemical Name or Material WYE-687
CAS 1062161-90-3
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C28H32N8O3
Quantity 50 mg
Solubility Information DMSO : 20.83 mg/mL (39.41 mM; ultrasonic and warming and heat to 80°C)
SMILES O=C(OC)NC1=CC=C(C2=NC(N3CCOCC3)=C4C(N(C5CCN(CC6=CC=CN=C6)CC5)N=C4)=N2)C=C1
Molecular Weight (g/mol) 528.61
Formula Weight 528.61
Percent Purity 98.1%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.