Learn More
MedChemExpress WST-8
Click to view available options
Quantity:
25 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
200mg
25mg
50mg
Specifications
Specifications
| Content And Storage | 4°C, sealed storage, away from moisture and light ∣ In solvent : -80°C, 6 months ∣ -20°C, 1 month (sealed storage, away from moisture and light) |
| Product Type | Dye Reagents |
| Quantity | 100 mg |
| Shipping Condition | Room temperature |
| Solubility | H2O : 83.33 mg/mL (138.54 mM; Need ultrasonic) ∣ DMSO : 10 mg/mL (16.63 mM; Need ultrasonic) |
| Target | HBV |
| Molecular Weight (g/mol) | 601.48 g/mol |
| CAS | 193149-74-5 |