Learn More
Brady™ WorkHorse™ Series Thermal Transfer Printable Labels, B-488
Product Code. 15685604
Change view
Click to view available options
Size (Metric):
19.05 x 6.35 mm (W x H)
22.86 x 22.86 mm (W x H)
50.8 x 6.35 mm (W x H)
Unit Size:
1 roll
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Artikelnummer. Size (Metric) unitSize
15685604 50.8 x 6.35 mm (W x H) 1 roll
13049302 22.86 x 22.86 mm (W x H) 1 roll
11553654 19.05 x 6.35 mm (W x H) 1 roll
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 15685604 Lieferant Brady™ Lieferanten-Nr. 029895
Nur noch null auf Lager
Zum Warenkorb hinzufügen
Zum Warenkorb hinzufügen
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Used to identify property from components to finished products. Brady™ WorkHorse™ Series Thermal Transfer Printable Labels are constructed of polyester and is used for thermal transfer printers.
  • Available in various sizes
Spezifikation
Autoclavable Autoclavable
Certifications/Compliance UL Recognized; CSA Accepted
Description Recommended ribbon series: R4300; R6200
Adhesive Type Acrylic, Permanent
Coating None
Color White
Finish Matte
Format Roll
Material Polyester (B-488)
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Thermal Transfer Printer Label
Sterility Non-sterile
For Use With (Application) Asset Tracking, Barcode Labeling, Circuit Board Identification - Through Hole Top, Component and Equipment Identification, General and Industrial Identification, Rating Plates
For Use With (Equipment) BBP81, i5100, i7100, PR Plus, IP Series, THT 76 mm Core Printers, i7500
Labels per Row 1
Min. Operating Temperature (Metric) -40°C
Model THT-15-488-2.5
No. per Pack 2500 Label(s) / Roll
Quantity 2500
Shape Rectangle
Size 50.80 mm (W) x 6.35 mm (H)
Size (Metric) 50.8 x 6.35 mm (W x H)
Temperature Range (English) -40°F to 320°F
Temperature Range (Metric) -40°C to +160°C
Thickness (English) 0.0036 in.
Thickness (Metric) 0.094 mm
Dimensions (LxW) 50.80 mm (W) x 6.35 mm (H)
Length (English) 0.25 in.
Width (English) 2 in.
Width (Metric) 50.8 mm
Width (English) Printable Area 50.80 mm
Height (English) 0.25 in.
Height (Metric) 6.35 mm
Alternate Part Number THT-15-488-2.5
Mehr anzeigen Weniger anzeigen