Learn More
Brady™ WorkHorse™ Series Thermal Transfer Printable Labels, B-488
Product Code. 13049302
Click to view available options
Size (Metric):
19.05 x 6.35 mm (W x H)
22.86 x 22.86 mm (W x H)
50.8 x 6.35 mm (W x H)
Unit Size:
1 roll
Product Code. 13049302
Brand: Brady™ 019261
Only null left
Add to basket
Add to basket
Used to identify property from components to finished products. Brady™ WorkHorse™ Series Thermal Transfer Printable Labels are constructed of polyester and is used for thermal transfer printers.
  • Available in various sizes
Specifications
Autoclavable Autoclavable
Certifications/Compliance UL Recognized; CSA Accepted
Description Recommended ribbon series: R4300; R6200
Adhesive Type Acrylic, Permanent
Coating None
Color White
Finish Matte
Format Roll
Material Polyester (B-488)
Printable Sides Single-side Printable
Print Technology Thermal Transfer
Product Type Thermal Transfer Printer Label
Sterility Non-sterile
For Use With (Application) Asset Tracking, Barcode Labeling, Circuit Board Identification - Through Hole Top, Component and Equipment Identification, General and Industrial Identification, Rating Plates
For Use With (Equipment) BBP81, i5100, i7100, PR Plus, IP Series, THT 76 mm Core Printers, i7500
Labels per Row 1
Min. Operating Temperature (Metric) -40°C
Model THT-164-488-1
No. per Pack 1000 Label(s) / Roll
Quantity 1000/Pk.
Shape Rectangle
Size 22.86 mm (W) x 22.86 mm (H)
Size (Metric) 22.86 x 22.86 mm (W x H)
Temperature Range (English) -40°F to 320°F
Temperature Range (Metric) -40°C to +160°C
Thickness (English) 0.0036 in.
Thickness (Metric) 0.091 mm
Vertical Repeat (Metric) 25.40 mm
Dimensions (LxW) 22.86 x 22.86 mm
Length (English) 0.9 in.
Width (English) 0.9 in.
Width (Metric) 22.86 mm
Width (English) Printable Area 22.86 mm
Height (English) 0.9 in.
Height (Metric) 22.86 mm
Alternate Part Number THT-164-488-1