Learn More
mb2 Wood Chair on Glides
Click to view available options
For Use With (Application):
For Table
Unit Size:
Each
Description
- Chair in wood
- Seat and back in 8 mm plywood
- Backrest adjustable in depth by wheel.
- Pneumatic cylinder middle
- 5-spoke base injected in nylon and fiberglass reinforced by a galvanized steel ring.
- Glides
Specifications
Specifications
| Product Type | Stool with Back |
| Color | Beech |
| Adjustable (Height) | Yes |
| Adjustable | Height |
| Autoclavable | Not Autoclavable |
| Back Style | Standard |
| For Use With (Application) | For Table |
| Height (Metric) | 700 mm |
| Quantity | 1 Ea. |
| Seating Height | 460 to 640 mm |
| Show More |