Learn More
Cofra Women's Safety Shoes, Paula Series
Click to view available options
Size:
2
3
4
6
7
Unit Size:
1 pair
Description
- Lace-up general purpose safety shoes
- Water-repellant breathable black nubuck uppers
- Steel toecaps resistant to 200J
- Tough EVA-nitrile anti-static footbed
- Breathable polyamide internal lining
- Slip-resistant polyurethane soles
Specifications
Specifications
| Closure Type | Lace-up |
| Color | Black |
| Footbed Material | EVA-Nitrile |
| Liner Material | Polyamide |
| Material | Leather |
| Size | 7 |
| Sole Features | Anti-slip |
| Sole Material | Polyurethane |
| Toe Material | Steel |
| Certifications/Compliance (Europe) | EN ISO 20345:2011 |
| Show More |