missing translation for 'onlineSavingsMsg'
Learn More

Wnt-C59, MedChemExpress

Product Code. 16472807
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16472807 50 mg 50mg
16469277 5 mg 5mg
16456728 10 mg 10mg
16456866 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16472807 Supplier Medchem Express Supplier No. HY15659

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Wnt-C59 (C59) is a highly potent and oral porcupine (PORCN) inhibitor with an IC50 of 74 pM.

Wnt-C59 (C59) is a highly potent and oral porcupine (PORCN) inhibitor with an IC50 of 74 pM.

Specifications

Chemical Name or Material Wnt-C59
CAS 1243243-89-1
Color White
Purity Grade Notes Research
Molecular Formula C25H21N3O
Quantity 50 mg
Synonym C59
Solubility Information DMSO : 50 mg/mL (131.77 mM; Need ultrasonic)
SMILES O=C(NC1=CC=C(C2=CC=CN=C2)C=C1)CC3=CC=C(C4=CC(C)=NC=C4)C=C3
Molecular Weight (g/mol) 379.45
Formula Weight 379.45
Percent Purity 99.32%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.